Learn More
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Product Code. 16446380 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black
Blue, Gray
Green, Black
Green, Clear
Yellow, Black
Lens Tint:
Amber
Clear
Gray
Light Gray, Indoor/Outdoor
Orange, Mirror
Vermillion
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Color Lens Tint Unit Size
16446380 Blue, Gray Gray Each
16416380 Green, Clear Clear Each
16426380 Blue, Gray Clear Each
16466380 Blue, Gray Light Gray, Indoor/Outdoor Each
16486380 Black Orange, Mirror Each
16476380 Yellow, Black Amber Each
16436380 Green, Black Clear Each
16406380 Black Vermillion Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16446380 Fornitore 3M™ N. del fornitore 7100185278
  • ONLINE EXCLUSIVE PRICE
    £18.00
    £11.25 / Pack of 1
    Save £6.75
    37% Off
Estimated Shipment: 22-02-2027
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
3M™ SecureFit™ 400X Safety Glasses have 3M™ Pressure Diffusion Temple Technology, flexible temple arms and combined soft nose bridge/brow guard. Lightweight anti-scratch & superior anti-fog, rimless polycarbonate lens for indoor/outdoor use.
  • Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
  • Wraparound design to improve the security of fit and prevent slipping
  • Padded temple touchpoints for increased comfort over the ears
  • Soft nose bridge for a more comfortable fit
  • Contoured brow guard provides additional coverage from flying particles
  • Superior anti-fog and anti-scratch properties meets the K&N marking requirements of EN166
  • Frame marking: SF400 Series, 3M EN166 FT
TRUSTED_SUSTAINABILITY
Specifica
Certifications/Compliance CE Approved
Lens Coating Anti-fog
Lens Tint Gray
Product Type Protective Eyewear
Frame Color Blue, Gray
Temple Style Pressure Diffusion Temple Technology
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.