Learn More
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Description
- Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
- Wraparound design to improve the security of fit and prevent slipping
- Padded temple touchpoints for increased comfort over the ears
- Soft nose bridge for a more comfortable fit
- Contoured brow guard provides additional coverage from flying particles
- Superior anti-fog and anti-scratch properties meets the K&N marking requirements of EN166
- Frame marking: SF400 Series, 3M EN166 FT
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Lens Coating | Anti-fog |
| Lens Tint | Vermillion |
| Product Type | Protective Eyewear |
| Frame Color | Black |
| Temple Style | Pressure Diffusion Temple Technology |
| Lens Material | Polycarbonate |