Learn More
3M™ SecureFit™ 100 Safety Glasses
Description
- Economical robust 3M™ SecureFit™ Safety Glasses range
- 3M's innovative SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head
- Slim wrap around design to look better on smaller faces
- Economical Bulk package (5-pack) available (black frame only)
- Flat and flexible temple design combines well with 3M™ Protective Earmuffs
- Wraparound design improves the security of fit and prevents slipping
- Easy to clean
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Certifications/Compliance (Europe) | EN 166:2001 |
| Lens Tint | Gray |
| Product Type | Protective Eyewear |
| Frame Color | Blue |
| Model | SF102AF-BLU-EU |
| Frame Material | Polycarbonate |
| UV Protection | UV400 |