missing translation for 'onlineSavingsMsg'
Learn More

seco Everolimus B (~90%), TRC

Product Code. 30380204
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380204 1 mg
30373678 0.25 mg
30386329 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380204 Supplier Toronto Research Chemicals Supplier No. TRCE9454101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769905-89-7
Molecular Formula C53H85NO15
Molecular Weight (g/mol) 976.24
Synonym (2S)-1-[2-[(2R,3R,6S)-6-[(2S,3E,5E,7E,9S,11R,13R,14R,15E,17R,20S,21R)-14,20-Dihydroxy-22-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]-2,13-dimethoxy-3,9,11,15,17,21-hexamethyl-12,18-dioxo-3,5,7,15-docosatetraen-1-yl]tetrahydro-2-hydroxy-3-methyl-2H-pyran-2-yl]-2-oxoacetyl]-2-piperidinecarboxylic Acid
SMILES CO[C@@H](C[C@@H]1CC[C@@H](C)[C@@](O)(O1)C(=O)C(=O)N2CCCC[C@H]2C(=O)O)\C(=C\C=C\C=C\[C@@H](C)C[C@@H](C)C(=O)[C@H](OC)[C@H](O)\C(=C\[C@@H](C)C(=O)C[C@H](O)[C@H](C)C[C@@H]3CC[C@@H](OCCO)[C@@H](C3)OC)\C)\C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C53H85NO15/c1-32(16-12-11-13-17-33(2)45(65-8)30-40-21-19-38(7)53(64,69-40)50(60)51(61)54-23-15-14-18-41(54)52(62)63)26-36(5)47(58)49(67-10)48(59)37(6)27-34(3)42(56)31-43(57)35(4)28-39-20-22-44(68-25-24-55)46(29-39)66-9/h11-13,16-17,27,32,34-36,38-41,43-46,48-49,55,57,59,64H,14-15,18-26,28-31H2,1-10H3,(H,62,63)/b13-11+,16-12+,33-17+,37-27+/t32-,34-,35-,36-,38-,39+,40+,41+,43+,44-,45+,46-,48-,49+,53-/m1/s1
Chemical Name or Material seco Everolimus B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.