CAS RN 769905-89-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
seco Everolimus B (~90%), TRC
CAS: 769905-89-7 Molecular Formula: C53H85NO15 Molecular Weight (g/mol): 976.24 Synonym: (2S)-1-[2-[(2R,3R,6S)-6-[(2S,3E,5E,7E,9S,11R,13R,14R,15E,17R,20S,21R)-14,20-Dihydroxy-22-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]-2,13-dimethoxy-3,9,11,15,17,21-hexamethyl-12,18-dioxo-3,5,7,15-docosatetraen-1-yl]tetrahydro-2-hydroxy-3-methyl-2H-pyran-2-yl]-2-oxoacetyl]-2-piperidinecarboxylic Acid SMILES: CO[C@@H](C[C@@H]1CC[C@@H](C)[C@@](O)(O1)C(=O)C(=O)N2CCCC[C@H]2C(=O)O)\C(=C\C=C\C=C\[C@@H](C)C[C@@H](C)C(=O)[C@H](OC)[C@H](O)\C(=C\[C@@H](C)C(=O)C[C@H](O)[C@H](C)C[C@@H]3CC[C@@H](OCCO)[C@@H](C3)OC)\C)\C