missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC61A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £366.00
Specifications
| Antigen | SEC61A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625310
|
Novus Biologicals
NBP2-93597-0.02ml |
0.02 mL |
£153.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605510
|
Novus Biologicals
NBP2-93597-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEC61A Polyclonal antibody specifically detects SEC61A in Mouse, Rat samples. It is validated for Western BlotSpecifications
| SEC61A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS (pH 7.3), 50% glycerol | |
| 29927 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| HSEC61, protein transport protein SEC61 alpha subunit, protein transport protein Sec61 subunit alpha, protein transport protein Sec61 subunit alpha isoform 1, SEC61, Sec61 alpha 1 subunit (S. cerevisiae), Sec61 alpha-1, sec61 homolog, SEC61A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-420 of human SEC61A1 (NP_037468.1). ARFSGNLLVSLLGTWSDTSSGGPARAYPVGGLCYYLSPPESFGSVLEDPVHAVVYIVFMLGSCAFFSKTWIEVSGSSAKDVAKQLKEQQMVMRGHRETSMVHELNRYIPTA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title