missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC61A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93597-0.1ml
This item is not returnable.
View return policy
Description
SEC61A Polyclonal antibody specifically detects SEC61A in Mouse, Rat samples. It is validated for Western Blot
Specifications
| SEC61A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HSEC61, protein transport protein SEC61 alpha subunit, protein transport protein Sec61 subunit alpha, protein transport protein Sec61 subunit alpha isoform 1, SEC61, Sec61 alpha 1 subunit (S. cerevisiae), Sec61 alpha-1, sec61 homolog, SEC61A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-420 of human SEC61A1 (NP_037468.1). ARFSGNLLVSLLGTWSDTSSGGPARAYPVGGLCYYLSPPESFGSVLEDPVHAVVYIVFMLGSCAFFSKTWIEVSGSSAKDVAKQLKEQQMVMRGHRETSMVHELNRYIPTA | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 29927 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction