missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC6 Antibody (9H5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-97500-0.025MG
This item is not returnable.
View return policy
Description
SEC6 Monoclonal specifically detects SEC6 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
| SEC6 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| EXOC3 | |
| Recombinant rat Sec6. | |
| 0.025 mg | |
| Primary | |
| Chicken, Mouse, Sheep, Rabbit, Pig, Monkey, Rat, Human, Canine, Bovine, Hamster | |
| Purified |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry | |
| 9H5 | |
| Western Blot 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoblotting | |
| exocyst complex component 3, Exocyst complex component Sec6, Sec 6 homolog, SEC6, SEC6L1, SEC6-like 1, SEC6-like 1 (S. cerevisiae), Sec6p | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 11336 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction