missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00 - £396.00
Specifications
| Antigen | SEC22C |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18219094
|
Novus Biologicals
NBP1-30760-0.1MG |
0.1 mg |
£133.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18219780
|
Novus Biologicals
NBP1-30760 |
0.5 mg |
£396.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEC22C Polyclonal specifically detects SEC22C in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SEC22C | |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| DKFZp761F2321, MGC13261, MGC5373, SEC22 vesicle trafficking protein homolog C (S. cerevisiae), SEC22 vesicle trafficking protein-like 3, SEC22 vesicle trafficking protein-like 3 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog C, SEC22 vesicle-trafficking protein-like 3, SEC22L3, secretion deficient 22C, vesicle-trafficking protein SEC22c | |
| SEC22C | |
| Primary | |
| Appears to be specific for SEC22C. |
| LYOPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| Sterilized water with No Preservative | |
| 9117 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title