missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-30760-0.1MG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SEC22C Polyclonal specifically detects SEC22C in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
| SEC22C | |
| Polyclonal | |
| Unconjugated | |
| Sterilized water with No Preservative | |
| SEC22C | |
| A synthetic from the c-terminal of human Vesicle trafficking protein SEC22C (SEC22L3) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in mouse and shares 95% identity with rat sequence. | |
| 0.1 mg | |
| Primary | |
| Appears to be specific for SEC22C. | |
| Mouse, Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| LYOPH | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| DKFZp761F2321, MGC13261, MGC5373, SEC22 vesicle trafficking protein homolog C (S. cerevisiae), SEC22 vesicle trafficking protein-like 3, SEC22 vesicle trafficking protein-like 3 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog C, SEC22 vesicle-trafficking protein-like 3, SEC22L3, secretion deficient 22C, vesicle-trafficking protein SEC22c | |
| Rabbit | |
| IgG purified | |
| RUO | |
| 9117 | |
| Reconstitute with 0.5 ml sterilized water. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido