missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£129.00 - £399.00
Specifications
| Antigen | SEC22B |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227904
|
Novus Biologicals
NB100-91277-0.1MG |
0.1 mg |
£129.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243930
|
Novus Biologicals
NB100-91277 |
0.5 mg |
£399.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEC22B Polyclonal specifically detects SEC22B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SEC22B | |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| ERS24, ERS-24sec22b, SEC22 vesicle trafficking protein homolog B (S. cerevisiae), SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene), SEC22 vesicle trafficking protein-like 1, SEC22 vesicle trafficking protein-like 1 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, SEC22, vesicle trafficking protein (S. cerevisiae)-like 1, SEC22L1ER-Golgi SNARE of 24 kDa, vesicle-trafficking protein SEC22b | |
| SEC22B | |
| Primary | |
| SEC22B. |
| LYOPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| O75396 | |
| 9554 | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title