missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-91277-0.1MG
This item is not returnable.
View return policy
Description
SEC22B Polyclonal specifically detects SEC22B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SEC22B | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| ERS24, ERS-24sec22b, SEC22 vesicle trafficking protein homolog B (S. cerevisiae), SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene), SEC22 vesicle trafficking protein-like 1, SEC22 vesicle trafficking protein-like 1 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, SEC22, vesicle trafficking protein (S. cerevisiae)-like 1, SEC22L1ER-Golgi SNARE of 24 kDa, vesicle-trafficking protein SEC22b | |
| Rabbit | |
| IgG purified | |
| RUO | |
| 9554 | |
| Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1-10 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| O75396 | |
| SEC22B | |
| A synthetic peptide as a part of human, rat, mouse, hamster, orangutan, chicken Vesicle-trafficking protein SEC22b (SEC22B, SEC22L1, ERS24) conjugated to an immunogenic carrier protein was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| SEC22B. | |
| Mouse, Rat, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction