missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC11C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79339
This item is not returnable.
View return policy
Description
SEC11C Polyclonal specifically detects SEC11C in Mouse samples. It is validated for Western Blot.
Specifications
| SEC11C | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.4, Microsomal signal peptidase 21 kDa subunit, SEC11 homolog C, SEC11 homolog C (S. cerevisiae), SEC11L3, SEC11-like protein 3, signal peptidase complex catalytic subunit SEC11C, SPase 21 kDa subunit, SPC21SEC11-like 3 (S. cerevisiae), SPCS4CSEC11-like 3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90701 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_079744 | |
| SEC11C | |
| The immunogen for this antibody is Sec11c. Peptide sequence VHRVIKVHEKDNGDIKFLTKGDNNEVDDRGLYKEGQNWLEKKDVVGRARG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Goat: 100%; Human: 100%; Rat: 100%; Xenopus: 92%; Chicken: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction