Learn More
Abcam SEAP Reporter Gene and Alkaline Phosphatase Activity Assay Kit (Luminometric)
Product Code. 32115721 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32115721 1 x 96 Tests 96 tests
1 options
Product Code. 32115721 Supplier Abcam Supplier No. ab3234601x96Tests
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure SEAP activity in 30m in Cell Samples, Cell/Tissue Extracts with SEAP Reporter Gene and Alkaline Phosphatase Activity Assay Kit AB323460. Sensitivity >0.0048 mU/well. Use with Microplate.
Measure SEAP activity in 30m in Cell Samples, Cell/Tissue Extracts with SEAP Reporter Gene and Alkaline Phosphatase Activity Assay Kit AB323460. Sensitivity >0.0048 mU/well. Use with Microplate.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 96 Tests