Learn More
Labconco™ Scroll Vacuum Pump Accessory, Silencer
Product Code. 17657835 Shop All Labconco Corporation Products
Click to view available options
For Use With:
Scroll Vacuum Pumps
Unit Size:
Each
Product Code. 17657835
Brand: Labconco™ 7587300
Only null left
Add to basket
Add to basket
For use with Labconco™ Scroll vacuum pump. Labconco™ Scroll Vacuum Pump Accessory, Silencer combines filtration efficiency with noise reduction.
Silencer for Labconco Scroll Pumps filters exhaust while silencing exhaust pump noise. It also prevents tip seal dust from contaminating the environment. Sound absorbent element is replaceable. Includes filter, centering ring and clamp to attach to exhaust outlet.
Specifications
Shipping Weight (English) 2.0 lb.
Shipping Weight (Metric) 0.9 kg
Includes Filter, Centering ring and Clamp to attach to exhaust outlet
For Use With Scroll Vacuum Pumps
Pump Type Vacuum Pump