missing translation for 'onlineSavingsMsg'
Learn More

Scribble Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£177.00 - £366.00

Specifications

Antigen Scribble
Dilution Western Blot 1:500 - 1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18612771
View Documents
Novus Biologicals
NBP2-93433-0.02ml
0.02 mL
£177.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18678280
View Documents
Novus Biologicals
NBP2-93433-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Scribble Polyclonal antibody specifically detects Scribble in Human samples. It is validated for Western Blot
Specifications

Specifications

Scribble
Western Blot
Unconjugated
Rabbit
Human
protein scribble homolog, CRIB1, SCRB1, SCRIB1, scribbled homolog (Drosophila), Vartul
Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1400 of human Scribble (NP_874365.3). ELCIQKAPGERLGISIRGGARGHAGNPRDPTDEGIFISKVSPTGAAGRDGRLRVGLRLLEVNQQSLLGLTHGEAVQLLRSVGDTLTVLVCDGFEASTDAALEVSPGVIANPFAAGIGHRNSLESISSIDRELSPEGPGKEKELPGQTLHWGPEATEAAGRGLQPLKLDYRALAAVPSAGSVQRVPSGAAGGKMAESPCSPSGQQPPSPPSPDELPANVKQAYRAFAAVPTSHPPEDAPAQPPTPGPAASPEQLSFRERQKYFELEVRVPQAEGPPKRVSLVGADDLRKMQEEEARKLQQKR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:1000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
23513
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.