missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scribble Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93433-0.02ml
This item is not returnable.
View return policy
Description
Scribble Polyclonal antibody specifically detects Scribble in Human samples. It is validated for Western Blot
Specifications
| Scribble | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| protein scribble homolog, CRIB1, SCRB1, SCRIB1, scribbled homolog (Drosophila), Vartul | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1400 of human Scribble (NP_874365.3). ELCIQKAPGERLGISIRGGARGHAGNPRDPTDEGIFISKVSPTGAAGRDGRLRVGLRLLEVNQQSLLGLTHGEAVQLLRSVGDTLTVLVCDGFEASTDAALEVSPGVIANPFAAGIGHRNSLESISSIDRELSPEGPGKEKELPGQTLHWGPEATEAAGRGLQPLKLDYRALAAVPSAGSVQRVPSGAAGGKMAESPCSPSGQQPPSPPSPDELPANVKQAYRAFAAVPTSHPPEDAPAQPPTPGPAASPEQLSFRERQKYFELEVRVPQAEGPPKRVSLVGADDLRKMQEEEARKLQQKR | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23513 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction