Learn More
JG Finneran Associates™ Screw Caps
Use these preassembled caps for a variety of applications. JG Finneran™ JG Finneran™ Screw Caps provide a positive physical lock to assure that preassembled components arrive intact.
Brand: JG Finneran Associates™ 5395F-09R
Code : 17
Additional Details : Weight : 0.01000kg
Description
- The patented Top Hat™ septa and closure provide a positive physical lock to assure that preassembled components arrive intact
- Preassembled R.A.M.™ Smooth Screw Thread Closure and Septa
- Designed to work in Agilent or other robotic arm autosamplers
- Available in a variety of colors
Specifications
9 mm | |
PTFE, Silicone | |
1000 Pk. | |
9 mm |
Red | |
PTFE/Silicone | |
Screw Cap | |
Agilent or Other Robotic Arm Autosamplers |