Learn More
JG Finneran Associates™ Screw Caps
Use these preassembled caps for a variety of applications. JG Finneran™ JG Finneran™ Screw Caps provide a positive physical lock to assure that preassembled components arrive intact.
£75.50 - £459.00
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
12403010
|
JG Finneran Associates™
5394F-09RB |
Royal Blue | PTFE, Rubber |
£275.00
Pack of 1000 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
12433010
|
JG Finneran Associates™
5394F-09R |
Red | PTFE, Rubber |
£427.00
Pack of 1000 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
12493010
|
JG Finneran Associates™
5395F-09R |
Red | PTFE, Silicone |
£287.00
Pack of 1000 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
12423020
|
JG Finneran Associates™
5396F-09RB |
Royal Blue | PTFE, Silicone |
£459.00
Pack of 1000 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10107080
|
JG Finneran Associates™
806550TH-08 |
Black | PTFE, Silicone |
£110.00
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
10700862
|
JG Finneran Associates™
806550TH-13 |
Black | PTFE, Silicone |
£75.50
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- The patented Top Hat™ septa and closure provide a positive physical lock to assure that preassembled components arrive intact
- Preassembled R.A.M.™ Smooth Screw Thread Closure and Septa
- Designed to work in Agilent or other robotic arm autosamplers
- Available in a variety of colors
Specifications
Screw Cap |