Learn More
DWK Life Sciences Screw Cap with Aperture, for safety standard ground sockets
For fixing the DURAN™ core in the DURAN™ threaded sleeve.
£2.12 - £82.70
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15951551
|
DWK Life Sciences
286215704 |
NS 45/40 |
£82.70
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15921551
|
DWK Life Sciences
286215207 |
NS 19/26 |
£18.40
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15911551
|
DWK Life Sciences
286215001 |
NS 14/23 |
£13.80
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15941551
|
DWK Life Sciences
286215404 |
NS 29/32 |
£30.85
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15844608
|
DWK Life Sciences
286215507 |
NS 34/35 |
£45.90
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10040151
|
DWK Life Sciences
286215301 |
NS 24/29 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The open screw cap reliably fixes the core of the DURAN™ security cuts in the sleeve. When the screw connection is opened, the ground joints are automatically separated from each other.
- Reliable fixation of the DURAN™ safety sections in one another (core and sleeve)
- When the screw connection is opened, the ground joints are automatically separated from each other
Specifications
10/Pk. |