Learn More
DWK Life Sciences Screw Cap with Aperture, for safety standard ground sockets
For fixing the DURAN™ core in the DURAN™ threaded sleeve.
Brand:  DWK Life Sciences 286215404
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 15941551
  • £30.85 / Pack of 10
Estimated Shipment: 09-07-2024
to see stock.
Add to basket
Description
Description
The open screw cap reliably fixes the core of the DURAN™ security cuts in the sleeve. When the screw connection is opened, the ground joints are automatically separated from each other.
  • Reliable fixation of the DURAN™ safety sections in one another (core and sleeve)
  • When the screw connection is opened, the ground joints are automatically separated from each other
Specifications
Specifications
10/Pk.
Suitable for fixing DURAN safety grinding
NS 29/32