Learn More
Agilent BioTek Scratch Assay App
GREENER_CHOICE
Product Code. 16562172 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Scratch Assay Starter Kit (1750012)
Unit Size:
Each
Product Code. 16562172
Brand: Agilent BioTek 1750011
Offers a simple interface for cell migration assay workflows, from kinetic capture through complete analysis. Pre-defined protocols for 24- and 96-well plates are available. Custom programming is quick and easy.
  • Offers a smooth, integrated workflow for kinetic image capture and powerful analysis of wound healing assays, performed with a Agilent BioTek Lionheart Automated Microscope or Cytation Cell Imaging Multi-Mode Reader
  • The Scratch Assay App and AutoScratch Wound Making Tool are critical components of the new Scratch Assay Starter Kit, which automates cell migration and wound healing assays
  • Simple installation and implementation
  • Pre-defined protocols for 24- and 96-well plate
  • Auto exposure and powerful image processing tools
  • Automatic statistics calculations: wound width, % confluence, maximum healing rate
  • For repeatable, consistent scratches and complete cell migration assay workflow consider the Scratch Assay Starter Kit
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Wound healing assays and Cell migration assays
For Use With (Equipment) Scratch Assay Starter Kit (1750012)
Type Software