Learn More
RETSCH Scrapers
Product Code. 10271505 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
For Mill GRINDOMIX GM 200
For Mill GRINDOMIX GM 300
For Mortar Grinder RM 200
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10271505 For Mill GRINDOMIX GM 300 Each
10094921 For Mill GRINDOMIX GM 200 Each
10036874 For Mortar Grinder RM 200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10271505 Supplier RETSCH Supplier No. 05.723.0002
Only null left
Add to basket
Add to basket
Specifications
Product Type Scraper
For Use With (Application) For Easier Recovery of Sticky Samples
For Use With (Equipment) For Mill GRINDOMIX GM 300