missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCN3B Recombinant Rabbit Monoclonal Antibody (5X9J0)
Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | SCN3B |
|---|---|
| Clone | 5X9J0 |
| Concentration | 1.28 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284755
|
Invitrogen™
MA557993 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30282602
|
Invitrogen™
MA557994 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: VCVEVPSETE AVQGNPMKLR CISCMKREEV EATTVVEWFY RPEGGKDFLI YEYRNGHQEV ESPFQGRLQW NGSKDLQDVS ITVLNVTLND SGLYTCNVSR EFEFEAHRPF VKTTRLIPLR V.
Voltage-gated sodium channels are transmembrane glycoprotein complexes composed of a large alpha subunit and one or more regulatory beta subunits. They are responsible for the generation and propagation of action potentials in neurons and muscle. This gene encodes one member of the sodium channel beta subunit gene family, and influences the inactivation kinetics of the sodium channel. Two alternatively spliced variants, encoding the same protein, have been identified.Specifications
| SCN3B | |
| 1.28 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9JK00, Q9NY72 | |
| 245956, 55800 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-145 of human SCN3B (NP_001035241.1). | |
| Antibody |
| 5X9J0 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| Scn3b | |
| 1110001K16Rik; 4833414B02Rik; ATFB16; brain and heart sodium channel beta 3 subunit; BRGDA7; HSA243396; KIAA1158; LOC540925 protein; Scn3b; Scnb3; sodium channel beta 3 subunit; sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta; sodium channel, voltage-gated, type III, beta subunit; sodium voltage-gated channel beta subunit 3; voltage-gated sodium channel beta-3 subunit | |
| Scn3b | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title