missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCN3B Recombinant Rabbit Monoclonal Antibody (5X9J0)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557994

 View more versions of this product

Product Code. 30282602

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VCVEVPSETE AVQGNPMKLR CISCMKREEV EATTVVEWFY RPEGGKDFLI YEYRNGHQEV ESPFQGRLQW NGSKDLQDVS ITVLNVTLND SGLYTCNVSR EFEFEAHRPF VKTTRLIPLR V.

Voltage-gated sodium channels are transmembrane glycoprotein complexes composed of a large alpha subunit and one or more regulatory beta subunits. They are responsible for the generation and propagation of action potentials in neurons and muscle. This gene encodes one member of the sodium channel beta subunit gene family, and influences the inactivation kinetics of the sodium channel. Two alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCN3B
Recombinant Monoclonal
1.28 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9JK00, Q9NY72
Scn3b
Recombinant fusion protein containing a sequence corresponding to amino acids 25-145 of human SCN3B (NP_001035241.1).
100 μL
Primary
Human, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
5X9J0
Unconjugated
Scn3b
1110001K16Rik; 4833414B02Rik; ATFB16; brain and heart sodium channel beta 3 subunit; BRGDA7; HSA243396; KIAA1158; LOC540925 protein; Scn3b; Scnb3; sodium channel beta 3 subunit; sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta; sodium channel, voltage-gated, type III, beta subunit; sodium voltage-gated channel beta subunit 3; voltage-gated sodium channel beta-3 subunit
Rabbit
Protein A
RUO
245956, 55800
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.