missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SCIN |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858753
|
Proteintech
11579-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848753
|
Proteintech
11579-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| SCIN | |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q60604, Q9Y6U3 | |
| 20259, 298975, 85477 | |
| SCIN Fusion Protein Ag2168 | |
| Primary | |
| -20°C | |
| SCIN |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Adseverin, KIAA1905, SCIN, scinderin | |
| SCIN | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title