missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11579-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| SCIN | |
| Polyclonal | |
| Unconjugated | |
| SCIN | |
| Adseverin, KIAA1905, SCIN, scinderin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20259, 298975, 85477 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q60604, Q9Y6U3 | |
| SCIN | |
| SCIN Fusion Protein Ag2168 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction