missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Schaedler Anaerobe KV Selective Agar with Lysed Horse Blood

Product Code. 10007222
This item is not returnable. View return policy
Product Code. 10007222 Supplier Thermo Scientific™ Supplier No. PO5020A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grow and isolate Gram negative bacteria with this highly nutritious, ready-to-use medium.

Grow and isolate Gram-negative bacteria, especially Bacteroides and Prevotella, with highly nutritious, ready-to-use Thermo Scientific™ Oxoid™ Schaedler Anaerobe KV Selective Agar with Lysed Horse Blood. The addition of kanamycin and vancomycin render the medium selective, while lysed horse blood supplies additional growth promoting substances.

Use Schaedler Anaerobe KV Selective Agar with Lysed Horse Blood for the detection and isolation of Gram-negative bacteria.

  • Selective: kanamycin and vancomycin render the medium selective.
  • Nutritious: lysed horse blood provides growth promoting factors to the medium.
  • Ready-to-use: convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Vemilion/ Peral copper
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 17g ± 5%
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.