missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | SCFD2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644765
|
Novus Biologicals
NBP2-38310-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116927
|
Novus Biologicals
NBP2-38310 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCFD2 Polyclonal specifically detects SCFD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SCFD2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ21060, FLJ39514, sec1 family domain containing 2, STXBP1L1sec1 family domain-containing protein 2, syntaxin binding protein 1-like 1, Syntaxin-binding protein 1-like 1 | |
| SCFD2 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8WU76 | |
| 152579 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ALAQVFCEESGLSPLLQKITDWDSSINLTFHKSKIAVDELFTSLRDIAGARSLLKQFKSVYVPGNHTHQASYKPLLKQVVEEIFHPERPDSVDIEH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title