missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38310
This item is not returnable.
View return policy
Description
SCFD2 Polyclonal specifically detects SCFD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SCFD2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q8WU76 | |
| SCFD2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ALAQVFCEESGLSPLLQKITDWDSSINLTFHKSKIAVDELFTSLRDIAGARSLLKQFKSVYVPGNHTHQASYKPLLKQVVEEIFHPERPDSVDIEH | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ21060, FLJ39514, sec1 family domain containing 2, STXBP1L1sec1 family domain-containing protein 2, syntaxin binding protein 1-like 1, Syntaxin-binding protein 1-like 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 152579 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction