missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £367.00
Specifications
| Antigen | SCD-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18210140
|
Novus Biologicals
NBP1-26393SS |
0.025 mL |
£159.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119619
|
Novus Biologicals
NBP1-26393 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| SCD-1 | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O00767 | |
| 6319 | |
| Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot# O00767] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
| SCD | |
| IgG | |
| Affinity Purified | |
| 41 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title