missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26393SS
This item is not returnable.
View return policy
Description
SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SCD-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
| Rabbit | |
| 41 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 | |
| O00767 | |
| SCD | |
| Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot# O00767] | |
| Affinity Purified | |
| RUO | |
| 6319 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction