missing translation for 'onlineSavingsMsg'
Learn More

SCAMP5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£155.00

Specifications

Antigen SCAMP5
Concentration 1 mg/mL
Dilution Western Blot 2 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18516472
View Documents
Novus Biologicals
NB120-3432-0.025mg
0.025 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

SCAMP5 Polyclonal specifically detects SCAMP5 in Rat samples. It is validated for Western Blot.
Specifications

Specifications

SCAMP5
Western Blot 2 μg/mL
Polyclonal
Rabbit
Membrane Trafficking and Chaperones
PBS with 0.05% Sodium Azide
192683
Synthetic Peptide: C N(217) Q P Q T Q Y S A T P N Y T Y S N(233)
Primary
Store at -20°C. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Rat
DKFZp686L1799, hSCAMP5, MGC24969, Sc5, secretory carrier membrane protein 5, secretory carrier-associated membrane protein 5
SCAMP5
IgG
Affinity Purified
NB120-3432 immunizing peptide corresponds to amino acid residues 217-233 from rat SCAMP 5. This sequence is 94% conserved in mouse.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.