missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3432-0.025mg
This item is not returnable.
View return policy
Description
SCAMP5 Polyclonal specifically detects SCAMP5 in Rat samples. It is validated for Western Blot.
Specifications
| SCAMP5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SCAMP5 | |
| Synthetic Peptide: C N(217) Q P Q T Q Y S A T P N Y T Y S N(233) | |
| 0.025 mg | |
| Membrane Trafficking and Chaperones | |
| 192683 | |
| Rat | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 μg/mL | |
| DKFZp686L1799, hSCAMP5, MGC24969, Sc5, secretory carrier membrane protein 5, secretory carrier-associated membrane protein 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB120-3432 immunizing peptide corresponds to amino acid residues 217-233 from rat SCAMP 5. This sequence is 94% conserved in mouse. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction