missing translation for 'onlineSavingsMsg'
Learn More

SCAMP2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NB120-3431-0.025mg

 View more versions of this product

Product Code. 18597441

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

SCAMP2 Polyclonal specifically detects SCAMP2 in Human, Mouse, Hamster samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCAMP2
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
SCAMP2
Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C
0.025 mg
Membrane Trafficking and Chaperones
10066
Human
IgG
Western Blot
1 mg/mL
Western Blot 2 μg/mL
secretory carrier membrane protein 2secretory carrier-associated membrane protein 2
Rabbit
Affinity Purified
RUO
Primary
Detects Secretory Carrier Membrane Protein 2 (SCAMP 2) from mouse testes and AtT20 cell extract, as well as Chinese hamster ovary (CHO) cell extract.
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only