missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£155.00
Specifications
| Antigen | SCAMP2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18597441
|
Novus Biologicals
NB120-3431-0.025mg |
0.025 mg | |||||||
Description
SCAMP2 Polyclonal specifically detects SCAMP2 in Human, Mouse, Hamster samples. It is validated for Western Blot.Specifications
| SCAMP2 | |
| Western Blot 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS with 0.05% Sodium Azide | |
| 10066 | |
| Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| secretory carrier membrane protein 2secretory carrier-associated membrane protein 2 | |
| SCAMP2 | |
| IgG | |
| Affinity Purified | |
| Detects Secretory Carrier Membrane Protein 2 (SCAMP 2) from mouse testes and AtT20 cell extract, as well as Chinese hamster ovary (CHO) cell extract. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title