missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sca1 (Ly-6A/E) Rat, anti-Mouse, APC, Clone: E13-161.7, STEMCELL Technologies
Rat Monoclonal Antibody
£143.10 - £456.30
Specifications
| Antigen | Sca1 (Ly-6A/E) |
|---|---|
| Clone | E13-161.7 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17118651
|
Stemcell Technologies
60032AZ.1 |
25 μg |
£143.10
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17108651
|
Stemcell Technologies
60032AZ |
100 μg |
£456.30
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family
- Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes
- Sca1 expression levels are strongly upregulated in all strains upon cellular activation
- Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells
- Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells
Specifications
| Sca1 (Ly-6A/E) | |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| APC | |
| Mouse | |
| Primary |
| E13-161.7 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title