Learn More
Sca1 (Ly-6A/E) Rat, anti-Mouse, APC, Clone: E13-161.7, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60032AZ
This item is not returnable.
View return policy
Description
- The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family
- Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes
- Sca1 expression levels are strongly upregulated in all strains upon cellular activation
- Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells
- Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells
Specifications
| Sca1 (Ly-6A/E) | |
| Monoclonal | |
| APC | |
| 100 μg | |
| Mouse | |
| Hematopoietic Stem and Progenitor Cells, Mammary Cells |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| E13-161.7 | |
| Rat | |
| Primary | |
| IgG2a κ |