missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 277011A dihydrochloride

Product Code. 18035632
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035632 10 mg 10mg
18044252 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18035632 Supplier Tocris Bioscience™ Supplier No. 4207/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective D3 antagonist

Selective dopamine D3 receptor antagonist (pKi values are 8.0, 6.0, 5.0 and <5.2 for D3, D2, 5-HT1D and 5-HT1B respectively). Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1226917-67-4
Molecular Formula C28H32Cl2N4O

Specifications

Quantity 10 mg
Formula Weight 511.49
Purity >98%
Molecular Weight (g/mol) 511.491
Chemical Name or Material SB 277011A dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.