accessibility menu, dialog, popup

UserName

CAS RN 1226917-67-4

NNHNONHClHCl

CAS RN 1226917-67-4

IUPAC Name: N-{4-[2-(6-cyano-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]cyclohexyl}quinoline-4-carboxamide dihydrochloride
Synonyms: N-{4-[2-(6-cyano-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]cyclohexyl}quinoline-4-carboxamide dihydrochloride
Molecular Weight (g/mol): 511.49
Molecular Formula: C28H32Cl2N4O
InChi Key: HEZIOTGUXSPDAK-UHFFFAOYSA-N
SMILES: Cl.Cl.O=C(NC1CCC(CCN2CCC3=C(C2)C=CC(=C3)C#N)CC1)C1=C2C=CC=CC2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results