missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 218078
GREENER_CHOICE

Product Code. 18054732
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054732 1 mg 1mg
18025192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054732 Supplier Tocris Bioscience™ Supplier No. 2560/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chk1 inhibitor

Inhibitor of checkpoint kinase 1 (Chk1) that displays selectivity over other protein kinases (IC50 values are 15, 250 and 1000 nM for Chk1, cdc2 and PKC respectively). Abrogates G2 cell cycle arrest caused by γ-irradiation and topoisomerase I inhibition. Potentiates cytotoxicity of DNA-damaging drugs, enhancing the efficacy of some chemotherapeutics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135897-06-2
Molecular Formula C24H15N3O3

Specifications

Quantity 1 mg
Formula Weight 393.39
Purity >98%
Molecular Weight (g/mol) 393.402
Chemical Name or Material SB 218078
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.