accessibility menu, dialog, popup

UserName

CAS RN 135897-06-2

NNOOHNO

CAS RN 135897-06-2

IUPAC Name: 28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaene-3,5-dione
Synonyms: 28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaene-3,5-dione
Molecular Weight (g/mol): 393.4
Molecular Formula: C24H15N3O3
InChi Key: OTPNDVKVEAIXTI-UHFFFAOYNA-N
SMILES: O=C1NC(=O)C2=C1C1=C3N(C4CCC(O4)N4C5=C(C=CC=C5)C2=C34)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results