missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SB 203580, 98%

Product Code. p-7082277
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 15464079

Brand: Thermo Scientific Chemicals J61482.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | CAS: 152121-47-6 | C21H16FN3OS, C21H16FN3OS | 377.44 g/mol

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152121-47-6
Molecular Formula C21H16FN3OS, C21H16FN3OS
Synonym 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole

Specifications

Quantity 25 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Highly specific new inhibitor of p38 kinase and MAP Kinase homologues
Formula Weight 377.44g/mol
Melting Point 235°C to 247°C
Product Type Signal Transduction Reagent
Form Powder
InChI Key CDMGBJANTYXAIV-UHFFFAOYNA-N
SMILES CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
IUPAC Name 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine
Molecular Weight (g/mol) 377.44
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.