missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivapore™ 5 Static Concentrators
Designed for quick and affordable concentration. Sartorius Vivapore 5 Static Concentrators are ready to use, disposable ultrafiltration devices for samples up to 5mL. These simple benchtop units are ideal for general purpose laboratory concentration or purification prior to further analysis. Available in packages of 4, 30 or 100.
£41.05 - £508.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | PES |
MWCO | 7.5 kDa |
Filtration Area | 20 cm2 |
Housing Material | Styrene acrylonitrile (SAN) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10448362
|
Sartorius
VP0503 |
4/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10120661
|
Sartorius
VP0501 |
30/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10797841
|
Sartorius
VP0502 |
100/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Economic and user-friendly- No need for additional equipment, pressure or a vacuum source
- Individual devices with disposable plastic stands for up to four units
- Simple concentration technique:
- Just fill the unit with the solution to be concentrated
- Wait for the desired concentration level to be achieved
- Then pipet the concentrated sample from the dead-stop reservoir
- Separate concentration chambers eliminate the risk of cross-contamination
- Each unit features a high-flow, 7.5 kDa PES membrane
- Ideal for samples that are sensitive to shearing forces or to pressure-induced stress
- Well suited for samples that require processing in a cold room environment
- Perfect for a variety of clinical and general research applications
- Urine concentration and subsequent analysis of Bence-Jones proteins by gel electrophoresis
- Concentrating macromolecules in cerebrospinal fluid (CSF) or other biological solutions
Specifications
Single-use | |
7.5 kDa | |
Styrene acrylonitrile (SAN) | |
82 mm |
PES | |
20 cm2 | |
1 to 5 mL | |
42 mm |