Learn More
Sartorius Pump Collar for Vivapure™ Miniprep and Maxiprep Protein Purification Spin Columns
Shop All Sartorius Products
Click to view available options
Includes:
Sealing cap
Unit Size:
Each
Description
- Use for processing large samples (greater than 20mL)
- Comes with sealing cap
Specifications
Specifications
| For Use With | Vivapure™ Miniprep and Maxiprep Protein Purification Spin Columns |
| Includes | Sealing cap |