missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Biosart™ 100 Nutrient Media

Product Code. 10307601 Shop All Sartorius Products
Change view
Click to view available options
Detects:
Acid-tolerant microorganisms
Enterococci
Escherichia coli and Coliforms
Pseudomonas aeruginosa
Total count
Yeasts and molds
Yeasts and molds and bacteria
Media Type:
Caso
Cetrimide
KF Strep ; Azide
Lauryl Sulphate/Teepol
Orange Serum
R2A
Sabouraud
TGE ; Total Count
Tergitol TTC
Total Count TTC
WL Differential ; Wallerstein Differentia
WL Media ; Wallerstein
Wort
m Endo
m-Green yeast and mold selective
m-Green yeast and mold, Schaufus Pottinger
mFC
Packaging Type:
Ampule
Quantity:
2.5 mL
Unit Size:
Pack of 50
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detects Media Type Packaging Type Quantity unitSize
10307601 Total count R2A Ampule 2.5 mL Pack of 50
11776025 Escherichia coli and Coliforms Lauryl Sulphate/Teepol Ampule 2.5 mL Pack of 50
10516281 Escherichia coli and Coliforms Tergitol TTC Ampule 2.5 mL Pack of 50
11756025 Total count Caso Ampule 2.5 mL Pack of 50
10747081 Pseudomonas aeruginosa Cetrimide Ampule 2.5 mL Pack of 50
10042301 Enterococci KF Strep ; Azide Ampule 2.5 mL Pack of 50
11766025 Yeasts and molds m-Green yeast and mold selective Ampule 2.5 mL Pack of 50
11753693 Escherichia coli and Coliforms m Endo Ampule 2.5 mL Pack of 50
10072301 Escherichia coli and Coliforms mFC Ampule 2.5 mL Pack of 50
10652602 Yeasts and molds m-Green yeast and mold, Schaufus Pottinger Ampule 2.5 mL Pack of 50
10544201 Acid-tolerant microorganisms Orange Serum Ampule 2.5 mL Pack of 50
11786025 Yeasts and molds Sabouraud Ampule 2.5 mL Pack of 50
10265421 Enterococci WL Differential ; Wallerstein Differentia Ampule 2.5 mL Pack of 50
11762335 Total count Total Count TTC Ampule 2.5 mL Pack of 50
15932530 Yeasts and molds Wort Ampule 2.5 mL Pack of 50
10178572 Yeasts and molds and bacteria WL Media ; Wallerstein Ampule 2.5 mL Pack of 50
10009920 Total count TGE ; Total Count Ampule 2.5 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 10307601 Supplier Sartorius Supplier No. 1640002RAK

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Microbiological Nutrient Media for Filtration Units

Biosart™ 100 Monitors have been specifically designed for microbiological testing of pharmaceuticals, food, beverages, water and other liquids. These sterile monitors with an incorporated membrane filter and cellulose pad are ready to use. Biosart™ 100 Nutrient Media for wetting the pad are available in individually sterilized, convenient plastic ampoules.Each box of Biosart™ 100 Nutrient Media contains 50 ampoules with sterile media, each with 2.5 ml and a lot certificate. If stored under proper conditions (+4°C), the culture media have a shelf life of 12 month (except for Endo, KF Strep, Lauryl Sulfate and Tergitol which have a 9-month shelf life). Biosart™ 100 Nutrient Media comply with international regulations and recommendations: International pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, guidelines of the food and beverage industries.Within the scope of the quality assurance procedure and the stringent quality control standards every batch has passed Sartorius in-house tests of growth promotion, sterility, physical and technical parameters have been passed successfully. Biosart™100 Nutrient Media are convenient in use and eliminating the handling of glass ampoules.

  • presterilized media
  • certificate of quality for every batch
  • in compliance with international standards
  • consistent recovery
  • Ready-to-use
  • long shelf life
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN, ISO
Content And Storage 4°C
Detects Total count
Form Liquid
Manufacturing Origin United States
Media Type R2A
Product Type Tubed Media
Sterility Sterile
Test Type Microbiological Tests
For Use With (Application) Microbiological testing of pharmaceuticals, food, beverages, Water and other liquids
Packaging Type Ampule
Quantity 2.5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.