missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Sartopore™ Liquid Filter Capsules
Ensures highly economic and safe filtration performance. Sartorius™ Sartopore™ Liquid Filter Capsules provide high-performance polyethersulfone (PES) liquid filtration from single-layer high flow solutions to specific Mycoplasma retentive filters. Available in a broad variety of different pore size combinations.
£837.40
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ISO 9001, USP Class VI |
Material (Membrane) | Double Layer PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
Product Code | Brand | Autoclavable | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Autoclavable | Price | Quantity & Availability | |||||
12333866
|
Sartorius™
5441307H5--OO--B |
Autoclavable |
£837.40
Pack of 5 |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
High-performance polyethersulfone (PES) liquid filters ensure highly economic and safe filtration performance. From single-layer high flow solutions to specific Mycoplasma retentive filters, Sartopore™2 perfectly adapts to your specific process needs. Available in a broad variety of different pore size combinationsSpecifications
Autoclavable | |
Double Layer PES | |
Sterile | |
For filtration of biologicals, pharmaceuticals, cell culture media and components and buffers | |
6.35mm Multiple Stepped Hose Barb | |
5Pack | |
107 mm |
ISO 9001, USP Class VI | |
0.2 μm | |
0.03 m2 | |
Polypropylene | |
6.35mm Multiple Stepped Hose Barb | |
Filter Capsule |