missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sartopore™ Liquid Filter Capsules

Ensures highly economic and safe filtration performance. Sartorius™ Sartopore™ Liquid Filter Capsules provide high-performance polyethersulfone (PES) liquid filtration from single-layer high flow solutions to specific Mycoplasma retentive filters. Available in a broad variety of different pore size combinations.

Brand:  Sartorius™ 5441307H5--OO--B

Code : 04

Additional Details : Weight : 0.34920kg

 View more versions of this product

Product Code. 12333866

  • £837.40 / Pack of 5
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High-performance polyethersulfone (PES) liquid filters ensure highly economic and safe filtration performance. From single-layer high flow solutions to specific Mycoplasma retentive filters, Sartopore™2 perfectly adapts to your specific process needs. Available in a broad variety of different pore size combinations
Specifications

Specifications

Autoclavable
Double Layer PES
Sterile
For filtration of biologicals, pharmaceuticals, cell culture media and components and buffers
6.35mm Multiple Stepped Hose Barb
5Pack
107 mm
ISO 9001, USP Class VI
0.2 μm
0.03 m2
Polypropylene
6.35mm Multiple Stepped Hose Barb
Filter Capsule
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers