missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartoclear Dynamics™: Lab V

Product Code. 15975830 Shop All Sartorius Products
Click to view available options
Bed Volume:
1,000mL
150mL
250mL
500mL
Quantity:
12 x 150mL Filter Units + 2g Filter Aid
12 x 1L Filter Units + 20g Filter Aid
12 x 1L Filter Units + 40g Filter Aid
12 x 500 mL Filter Units + 20g Filter Aid
24 x 150mL Filter Units + 5g Filter Aid
24 x 1L Filter Units + 10g Filter Aid
24 x 250mL Filter Units + 10g Filter Aid
24 x 500mL Filter Units + 10g Filter Aid
24 x 500mL Filter Units + 5g Filter Aid
Unit Size:
Pack of 12
Pack of 24
This item is not returnable. View return policy

Product Code. 15975830

Brand: Sartorius SDLV015005C2

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For simultaneous clarification and sterile filtration of 50 mL to 1 L mammalian cell cultures in one step without centrifugation.

Sartoclear Dynamics™ Lab V kits are designed for simultaneous clarification and sterile filtration of 50 mL to 1 L of mammalian cell culture broth to prepare samples for subsequent downstream processing steps, such as protein purification and concentration. The convenient kits combine pouches of filter aid for clarification and vacuum filtration units for sterile filtration.

The Sartoclear Dynamics™ Lab V Kits combine the use of a filter aid for clarification and a vacuum filtration unit with a 0.22 μm polyethersulfone membrane for sterile filtration of up to 1 L mammalian cell cultures. The ready-to-use filter aid is supplied in dust-free and gamma-irradiated pouches.

The Sartolab™ RF bottletop filters with receiver flasks, sterilized by gamma irradiation are
available in volume capacities ranging from 150 mL to 1,000 mL.

  • Clarify high-density mammalian cell culture in minutes
  • Out-of-the-box solution
  • Fast and effortless
  • No need for centrifugation

Ten kits are available to meet all your needs based on your
filtration volume and the cell density of your mammalian
cell cultures.

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Diatomaceous Earth/Heavy metal-free polystyrene/Heavy metal-free polyethylene
Product Type Sartoclear Dynamics&trade: Lab V kits have been designed for cell harvesting of mammalian cell cultures, such as CHO, HEK, hybridomas and many others, with high cell densities.
Bed Volume 150mL
Quantity 24 x 150mL Filter Units + 5g Filter Aid
Flow Rate 0.22 μm polyethersulfone/high flow rate
Includes Each Sartoclear Dynamics&trade: Lab V kit is comprised of pouches of filter aid and vacuum filtration units (Sartolab&trade: RF) that match your needs. Find the right kit in just two easy steps: 1. Determine the volume range of your sample to be filtered. 2. Then reference this range to the cell density of your cell culture
Content And Storage ambient
For Use With (Application) mammalian cell harvesting/culture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.