missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars E2 Antibody (4A6C9), Novus Biologicals™
Mouse Monoclonal Antibody
£104.00
Specifications
| Antigen | Sars E2 |
|---|---|
| Clone | 4A6C9 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry-Paraffin |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sars E2 Monoclonal specifically detects Sars E2 in SARS-CoV samples. It is validated for Western Blot, ELISA.Specifications
| Sars E2 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Supernatant | |
| Virus | |
| Purified recombinant fragment of SARS-E2 glycoprotein precursor expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 4A6C9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Subclonal supernatant with No Preservative | |
| IgG1 | |
| Tissue culture supernatant | |
| Specific for SARS-E2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title